Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.6370 in Dia, 0.000200 in Tolerance, Steel
Catalog No. 5000530902
Encompass
This is a Class ZZ Plus Plug Gage designed for precise measurement applications. It features a 0.6370 inch diameter and a tight 0.000200 inch tolerance, ensuring high accuracy. Constructed from durable steel, it is suitable for various industrial and laboratory uses.
  • Class ZZ Plus plug gage
  • 0.6370 inch diameter
  • 0.000200 inch tolerance
  • Made from steel material
  • Ideal for precise measurement tasks
Catalog No. 50-005-30902 Supplier MSC Supplier No. 78388220
This is a Class ZZ Plus Plug Gage designed for precise measurement applications. It features a 0.6370 inch diameter and a tight 0.000200 inch tolerance, ensuring high accuracy. Constructed from durable steel, it is suitable for various industrial and laboratory uses.
  • Class ZZ Plus plug gage
  • 0.6370 inch diameter
  • 0.000200 inch tolerance
  • Made from steel material
  • Ideal for precise measurement tasks