Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.6560 Inch Diameter, Black Oxide
Catalog No. 5000530521
Encompass
This Class ZZ Plus Plug Gage is engineered for precise measurement and inspection tasks. Featuring a 0.6560 inch diameter and a tight +0.000200 inch tolerance, it ensures accuracy in critical applications. The durable black oxide coating provides enhanced resistance to wear and corrosion.
  • Class ZZ Plus for high precision gaging.
  • Diameter of 0.6560 "
  • Tight tolerance of +0.000200 "
  • Black oxide finish for durability.
Catalog No. 50-005-30521 Supplier MSC Supplier No. 78388410
This Class ZZ Plus Plug Gage is engineered for precise measurement and inspection tasks. Featuring a 0.6560 inch diameter and a tight +0.000200 inch tolerance, it ensures accuracy in critical applications. The durable black oxide coating provides enhanced resistance to wear and corrosion.
  • Class ZZ Plus for high precision gaging.
  • Diameter of 0.6560 "
  • Tight tolerance of +0.000200 "
  • Black oxide finish for durability.