Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.6620 in Diameter, Steel
Catalog No. 5000527271
Encompass
This plug gage is designed for precise measurement of internal diameters. Manufactured from steel, it provides reliable performance and accuracy for quality control applications.
  • Measures internal diameters with high precision
  • Constructed from durable steel for long-lasting use
  • Features a Class ZZ Plus designation for specific tolerance requirements
  • Maintains a tight tolerance of 0.000200 inch
Catalog No. 50-005-27271 Supplier MSC Supplier No. 78388477
Add to Cart
Add to Cart
This plug gage is designed for precise measurement of internal diameters. Manufactured from steel, it provides reliable performance and accuracy for quality control applications.
  • Measures internal diameters with high precision
  • Constructed from durable steel for long-lasting use
  • Features a Class ZZ Plus designation for specific tolerance requirements
  • Maintains a tight tolerance of 0.000200 inch