Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.6990 inch Diameter, Black Oxide
Catalog No. 5000527501
Encompass
The Class ZZ Plus Plug Gage is a precision measuring tool designed for accurate gauging. It features a 0.6990 inch diameter with a tight tolerance of +0.000200 inch and a black oxide coating. This plug gage is suitable for various inspection and quality control applications.
  • Tolerance class: Class ZZ Plus
  • Diameter of measurement: 0.6990 inch
  • Tolerance: +0.000200 inch
  • Coating/finish: Black Oxide
  • Country of origin: China
  • UNSPSC Code: 41111600
Catalog No. 50-005-27501 Supplier MSC Supplier No. 56240526
The Class ZZ Plus Plug Gage is a precision measuring tool designed for accurate gauging. It features a 0.6990 inch diameter with a tight tolerance of +0.000200 inch and a black oxide coating. This plug gage is suitable for various inspection and quality control applications.
  • Tolerance class: Class ZZ Plus
  • Diameter of measurement: 0.6990 inch
  • Tolerance: +0.000200 inch
  • Coating/finish: Black Oxide
  • Country of origin: China
  • UNSPSC Code: 41111600