Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.7100 in Dia, Steel
Catalog No. 5000528751
Encompass
The Class ZZ Plus Plug Gage is designed for precise measurement applications. It features a 0.7100 in diameter and a tolerance of 0.000200 in, making it suitable for checking bore sizes and other critical dimensions. Made from steel, this plug gage ensures durability and accuracy.
  • Diameter of 0.7100 in
  • Tolerance of 0.000200 in
  • Constructed from durable steel
Catalog No. 50-005-28751 Supplier MSC Supplier No. 78388956
The Class ZZ Plus Plug Gage is designed for precise measurement applications. It features a 0.7100 in diameter and a tolerance of 0.000200 in, making it suitable for checking bore sizes and other critical dimensions. Made from steel, this plug gage ensures durability and accuracy.
  • Diameter of 0.7100 in
  • Tolerance of 0.000200 in
  • Constructed from durable steel