Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.7450 in Dia, Steel
Catalog No. 5000510883
Encompass
This plug gage is designed for precise measurement applications, featuring a specific diameter and tolerance to ensure accuracy. Constructed from durable steel, it is suitable for various industrial and laboratory settings requiring high precision inspection.
  • Measures 0.7450 inch diameter
  • Provides a tolerance of 0.000200 inch
  • Constructed from steel for durability and precision
Catalog No. 50-005-10883 Supplier MSC Supplier No. 78389301
This plug gage is designed for precise measurement applications, featuring a specific diameter and tolerance to ensure accuracy. Constructed from durable steel, it is suitable for various industrial and laboratory settings requiring high precision inspection.
  • Measures 0.7450 inch diameter
  • Provides a tolerance of 0.000200 inch
  • Constructed from steel for durability and precision