Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.7750 in Diameter, Black Oxide
Catalog No. 5000515048
Encompass
This Class ZZ Plus plug gage is a precision tool designed for accurate measurement. It features a durable black oxide coating for enhanced longevity and resistance to wear, making it suitable for various industrial applications.
  • Class ZZ Plus for precise measurements.
  • Features a 0.7750 in diameter.
  • Provides a tolerance of +0.000200 in.
  • Includes a durable black oxide coating.
  • Manufactured in China.
Catalog No. 50-005-15048 Supplier MSC Supplier No. 56241300
This Class ZZ Plus plug gage is a precision tool designed for accurate measurement. It features a durable black oxide coating for enhanced longevity and resistance to wear, making it suitable for various industrial applications.
  • Class ZZ Plus for precise measurements.
  • Features a 0.7750 in diameter.
  • Provides a tolerance of +0.000200 in.
  • Includes a durable black oxide coating.
  • Manufactured in China.