Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.7760 in Dia, Black Oxide
Catalog No. 5000531285
Encompass
This plug gage is designed for precise measurement applications. It features a 0.7760 inch diameter with a tight tolerance of +0.000200 inch, ensuring accuracy for critical inspections. The black oxide finish provides durability and corrosion resistance.
  • Precision measurement for accurate inspections
  • 0.7760 inch diameter
  • Tight tolerance of +0.000200 inch
  • Black oxide coating for durability and corrosion resistance
Catalog No. 50-005-31285 Supplier MSC Supplier No. 56241318
This plug gage is designed for precise measurement applications. It features a 0.7760 inch diameter with a tight tolerance of +0.000200 inch, ensuring accuracy for critical inspections. The black oxide finish provides durability and corrosion resistance.
  • Precision measurement for accurate inspections
  • 0.7760 inch diameter
  • Tight tolerance of +0.000200 inch
  • Black oxide coating for durability and corrosion resistance