Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.787 in Dia, Black Oxide Finish
Catalog No. 5000527669
Encompass
This plug gage is a precision measuring tool designed for accurate diameter verification. Its 0.787 in diameter and tight tolerance of +0.000200 in (0.0002 mm) ensure high precision in measurements. The black oxide finish provides enhanced durability and corrosion resistance, extending the life of the gage. This tool is ideal for quality control and inspection tasks where exact measurements are critical.
  • Designed for accurate diameter verification.
  • Features a 0.787 in diameter and a tight tolerance of +0.000200 in (0.0002 mm) for high precision.
  • Black oxide finish provides enhanced durability and corrosion resistance.
  • Ideal for quality control and inspection tasks where exact measurements are critical.
Catalog No. 50-005-27669 Supplier MSC Supplier No. 56241425
This plug gage is a precision measuring tool designed for accurate diameter verification. Its 0.787 in diameter and tight tolerance of +0.000200 in (0.0002 mm) ensure high precision in measurements. The black oxide finish provides enhanced durability and corrosion resistance, extending the life of the gage. This tool is ideal for quality control and inspection tasks where exact measurements are critical.
  • Designed for accurate diameter verification.
  • Features a 0.787 in diameter and a tight tolerance of +0.000200 in (0.0002 mm) for high precision.
  • Black oxide finish provides enhanced durability and corrosion resistance.
  • Ideal for quality control and inspection tasks where exact measurements are critical.