Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.7970 in Dia, 0.000200 in Tolerance
Catalog No. 5000530527
Encompass
This plug gage is designed for precise measurement applications. It features a steel construction and is classified as ZZ Plus, providing a high degree of accuracy for quality control and inspection tasks.
  • Class ZZ Plus
  • Diameter of measurement 0.7970 in
  • Tolerance of 0.000200 in
  • Made from steel
Catalog No. 50-005-30527 Supplier MSC Supplier No. 78389822
This plug gage is designed for precise measurement applications. It features a steel construction and is classified as ZZ Plus, providing a high degree of accuracy for quality control and inspection tasks.
  • Class ZZ Plus
  • Diameter of measurement 0.7970 in
  • Tolerance of 0.000200 in
  • Made from steel