Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.8170 in., Black Oxide
Catalog No. 5000514522
Encompass
This Class ZZ Plus Plug Gage is designed for precision gaging, featuring a 0.8170 inch diameter and a tight +0.000200 inch tolerance. The black oxide finish enhances durability and provides corrosion resistance, ensuring reliable performance in industrial and laboratory settings.
  • Class ZZ Plus designation
  • 0.8170 inch diameter for accurate measurements
  • +0.000200 inch tolerance ensures high precision
  • Black oxide finish for durability and corrosion resistance
Catalog No. 50-005-14522 Supplier MSC Supplier No. 56241722
This Class ZZ Plus Plug Gage is designed for precision gaging, featuring a 0.8170 inch diameter and a tight +0.000200 inch tolerance. The black oxide finish enhances durability and provides corrosion resistance, ensuring reliable performance in industrial and laboratory settings.
  • Class ZZ Plus designation
  • 0.8170 inch diameter for accurate measurements
  • +0.000200 inch tolerance ensures high precision
  • Black oxide finish for durability and corrosion resistance