Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.879 in Diameter, Black Oxide Finish
Catalog No. 5000528848
Encompass
This plug gage is a Class ZZ Plus instrument designed for precise measurements. It features a 0.879 inch diameter and a black oxide finish, making it suitable for a variety of industrial and laboratory applications.
  • Class ZZ Plus design
  • Black oxide coating
  • 0.879 inch diameter measurement
  • +0.0002 inch tolerance
Catalog No. 50-005-28848 Supplier MSC Supplier No. 56242340
This plug gage is a Class ZZ Plus instrument designed for precise measurements. It features a 0.879 inch diameter and a black oxide finish, making it suitable for a variety of industrial and laboratory applications.
  • Class ZZ Plus design
  • Black oxide coating
  • 0.879 inch diameter measurement
  • +0.0002 inch tolerance