Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.8830 in Dia, Black Oxide
Catalog No. 5000530216
Encompass
This SPI Class ZZ Plus Plug Gage features a 0.8830 in diameter with a +0.000200 in tolerance. It is finished with black oxide for durability and precise measurement.
  • 0.8830 in diameter of measurement
  • +0.000200 in tolerance
  • Black oxide coating and finish
  • Made in China
Catalog No. 50-005-30216 Supplier MSC Supplier No. 56242381
Add to Cart
Add to Cart
This SPI Class ZZ Plus Plug Gage features a 0.8830 in diameter with a +0.000200 in tolerance. It is finished with black oxide for durability and precise measurement.
  • 0.8830 in diameter of measurement
  • +0.000200 in tolerance
  • Black oxide coating and finish
  • Made in China