Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.8840 in Diameter, Steel
Catalog No. 5000535787
Encompass
This plug gage is designed for precision measurement with a diameter of 0.8840 inches and a tolerance of 0.000200 inches. Made from durable steel, it is suitable for various industrial and scientific applications requiring accurate bore and hole measurement.
  • Precision measurement for bore and hole diameters
  • Constructed from durable steel for longevity
  • Class ZZ Plus for high accuracy
  • Ideal for quality control and inspection tasks
Catalog No. 50-005-35787 Supplier MSC Supplier No. 88988407
This plug gage is designed for precision measurement with a diameter of 0.8840 inches and a tolerance of 0.000200 inches. Made from durable steel, it is suitable for various industrial and scientific applications requiring accurate bore and hole measurement.
  • Precision measurement for bore and hole diameters
  • Constructed from durable steel for longevity
  • Class ZZ Plus for high accuracy
  • Ideal for quality control and inspection tasks