Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.8920 in, 0.0002 in Tolerance
Catalog No. 5000512818
Encompass
This Class ZZ Plus plug gage is designed for precise measurement and calibration. With a 0.8920 inch diameter and a 0.0002 inch tolerance, it is suitable for Go Inspection and for accurately setting various measuring instruments.
  • Class ZZ Plus
  • 0.8920 inch diameter
  • 0.0002 inch tolerance
  • Overall length of 2.0000 inches
  • Steel material
  • Conforms to ASME B89.1 standards
  • Includes a certificate of accuracy with NIST traceability
  • 10 micro coating/finish
  • Ideal for Go Inspection
  • Used for setting micrometers, verniers, and other measuring devices
Catalog No. 50-005-12818 Supplier MSC Supplier No. 88989207
This Class ZZ Plus plug gage is designed for precise measurement and calibration. With a 0.8920 inch diameter and a 0.0002 inch tolerance, it is suitable for Go Inspection and for accurately setting various measuring instruments.
  • Class ZZ Plus
  • 0.8920 inch diameter
  • 0.0002 inch tolerance
  • Overall length of 2.0000 inches
  • Steel material
  • Conforms to ASME B89.1 standards
  • Includes a certificate of accuracy with NIST traceability
  • 10 micro coating/finish
  • Ideal for Go Inspection
  • Used for setting micrometers, verniers, and other measuring devices