Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.8950 inch Diameter, Steel
Catalog No. 5000535917
Encompass
This plug gage is a Class ZZ Plus tool designed for precise measurement and inspection. Constructed from steel, it offers accurate gaging for various applications.
  • Class ZZ plus
  • 0.8950 inch diameter
  • 0.000200 inch tolerance
  • Steel material
  • Suitable for go inspection and setting measuring devices
Catalog No. 50-005-35917 Supplier MSC Supplier No. 78390804
This plug gage is a Class ZZ Plus tool designed for precise measurement and inspection. Constructed from steel, it offers accurate gaging for various applications.
  • Class ZZ plus
  • 0.8950 inch diameter
  • 0.000200 inch tolerance
  • Steel material
  • Suitable for go inspection and setting measuring devices