Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.9090 in Dia, Steel
Catalog No. 5000515619
Encompass
This precision plug gage is an essential measuring device for accurate 'Go' inspection and for calibrating various measuring instruments. Crafted from durable steel, it offers reliable performance for demanding applications. Each gage conforms to ASME B89.1 standards and includes a Certificate of Accuracy with NIST Traceability, ensuring verifiable precision.
  • Manufactured from high-quality steel.
  • Features a Class ZZ Plus tolerance.
  • Designed for "Go" inspection applications.
  • Suitable for setting micrometers, verniers, and other measuring devices.
  • Conforms to ASME B89.1 standards.
  • Includes a Certificate of Accuracy with NIST Traceability.
  • Coating/finish features 10 micro.
Catalog No. 50-005-15619 Supplier MSC Supplier No. 78390945
This precision plug gage is an essential measuring device for accurate 'Go' inspection and for calibrating various measuring instruments. Crafted from durable steel, it offers reliable performance for demanding applications. Each gage conforms to ASME B89.1 standards and includes a Certificate of Accuracy with NIST Traceability, ensuring verifiable precision.
  • Manufactured from high-quality steel.
  • Features a Class ZZ Plus tolerance.
  • Designed for "Go" inspection applications.
  • Suitable for setting micrometers, verniers, and other measuring devices.
  • Conforms to ASME B89.1 standards.
  • Includes a Certificate of Accuracy with NIST Traceability.
  • Coating/finish features 10 micro.