Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.9120 in Diameter, 0.000200 in Tolerance
Catalog No. 5000528938
Encompass
This Class ZZ Plus Plug Gage is a precision measuring tool designed for accurately verifying hole sizes. Constructed from durable steel, it ensures reliable and consistent performance in demanding applications.
  • Precisely measures with a 0.9120 inch diameter.
  • Maintains tight control with a 0.000200 inch tolerance.
  • Features a robust steel construction for longevity.
  • Suitable for various industrial measurement needs.
Catalog No. 50-005-28938 Supplier MSC Supplier No. 78390978
Add to Cart
Add to Cart
This Class ZZ Plus Plug Gage is a precision measuring tool designed for accurately verifying hole sizes. Constructed from durable steel, it ensures reliable and consistent performance in demanding applications.
  • Precisely measures with a 0.9120 inch diameter.
  • Maintains tight control with a 0.000200 inch tolerance.
  • Features a robust steel construction for longevity.
  • Suitable for various industrial measurement needs.