Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.9570 in Diameter, Steel
Catalog No. 5000531667
Encompass
This plug gage is a Class ZZ Plus precision measuring tool, constructed from steel, and designed for accurate diameter verification. It features a specific diameter and tolerance for precise fit and function.
  • Diameter of 0.9570 in
  • Tolerance of 0.000200 in
  • Made of steel
  • Class ZZ Plus for high precision
Catalog No. 50-005-31667 Supplier MSC Supplier No. 78391422
This plug gage is a Class ZZ Plus precision measuring tool, constructed from steel, and designed for accurate diameter verification. It features a specific diameter and tolerance for precise fit and function.
  • Diameter of 0.9570 in
  • Tolerance of 0.000200 in
  • Made of steel
  • Class ZZ Plus for high precision