Learn More
MSC SPI Plug Gage, Class ZZ Plus, 0.964 in Diameter, 0.0002 in Tolerance
Catalog No. 5000535991
Encompass
This plug gage is designed for precise measurement applications, offering high accuracy for checking bore sizes. It features a specific diameter and a tight tolerance, ensuring reliable performance in metrology tasks.
  • Diameter of measurement: 0.964 inch
  • Tolerance: 0.0002 inch
  • Made from steel for durability
Catalog No. 50-005-35991 Supplier MSC Supplier No. 78391497
This plug gage is designed for precise measurement applications, offering high accuracy for checking bore sizes. It features a specific diameter and a tight tolerance, ensuring reliable performance in metrology tasks.
  • Diameter of measurement: 0.964 inch
  • Tolerance: 0.0002 inch
  • Made from steel for durability