Learn More
MSC SPI Plug Gage Handle, 0.06-0.426 in, 6061 Aluminum, 0.5 x 0.75 in
Catalog No. 5000527444
Encompass
This plug gage handle is engineered for precise Go/No Go applications, supporting quality control and measurement tasks. It is fabricated from robust 6061 aluminum, providing durability and a reliable grip. The handle is designed to accommodate specific plug gage sizes, with clear indicators for straightforward operation.
  • Designed for Go and No Go applications
  • Accommodates plug gages with a measurement range of 0.06-0.426 inches
  • Constructed from 6061 aluminum
  • Measures 0.5 inch wide by 0.75 inch high
  • Go and No Go clearly marked for easy identification
Catalog No. 50-005-27444 Supplier MSC Supplier No. 67642439
This plug gage handle is engineered for precise Go/No Go applications, supporting quality control and measurement tasks. It is fabricated from robust 6061 aluminum, providing durability and a reliable grip. The handle is designed to accommodate specific plug gage sizes, with clear indicators for straightforward operation.
  • Designed for Go and No Go applications
  • Accommodates plug gages with a measurement range of 0.06-0.426 inches
  • Constructed from 6061 aluminum
  • Measures 0.5 inch wide by 0.75 inch high
  • Go and No Go clearly marked for easy identification