Learn More
MSC SPI Plug Gage Set, 0.201 to 0.3 in Range, Class ZZ, 100 Pc
Catalog No. 5000536634
Encompass
This plug gage set is designed for precise diameter measurement in industrial applications. Made from durable steel, it offers a measurement range of 0.201 to 0.3 inches with Class ZZ tolerance, ensuring high accuracy. The set includes 100 individual pieces, providing a comprehensive solution for various sizing needs.
  • Accurate diameter measurement
  • Features Class ZZ tolerance for high precision
  • Constructed from steel for durability
  • Measures diameters from 0.201 to 0.3 inches
  • Contains 100 pieces for a wide range of applications
  • Double-ended design
Catalog No. 50-005-36634 Supplier MSC Supplier No. 17135450
This plug gage set is designed for precise diameter measurement in industrial applications. Made from durable steel, it offers a measurement range of 0.201 to 0.3 inches with Class ZZ tolerance, ensuring high accuracy. The set includes 100 individual pieces, providing a comprehensive solution for various sizing needs.
  • Accurate diameter measurement
  • Features Class ZZ tolerance for high precision
  • Constructed from steel for durability
  • Measures diameters from 0.201 to 0.3 inches
  • Contains 100 pieces for a wide range of applications
  • Double-ended design