Learn More
MSC SPI Plug Gage Set, Class ZZ, 0.0197-0.0394 in Range, 51 Pc
Catalog No. 5000536318
Encompass
This plug gage set provides precise measurement for diameters within the specified range. It is designed for accuracy in various applications, ensuring reliable dimensional inspection.
  • Class ZZ plug gage set
  • Measurement range of 0.0197-0.0394 in
  • Includes 51 pieces for comprehensive coverage
  • Features a tolerance of ±0.0020 in
  • Constructed from durable steel
  • Designed with a double end type
Catalog No. 50-005-36318 Supplier MSC Supplier No. 17135666
This plug gage set provides precise measurement for diameters within the specified range. It is designed for accuracy in various applications, ensuring reliable dimensional inspection.
  • Class ZZ plug gage set
  • Measurement range of 0.0197-0.0394 in
  • Includes 51 pieces for comprehensive coverage
  • Features a tolerance of ±0.0020 in
  • Constructed from durable steel
  • Designed with a double end type