Learn More
MSC SPI Class ZZ Minus Plug Gage, Steel, 0.3600 in. Diameter, 1 ea.
Catalog No. 5000511703
Encompass
This Class ZZ minus plug and pin gage is designed for precise diameter measurements. Constructed from steel, it provides reliable performance in dimensional inspection and calibration applications.
  • Nominal diameter of 0.3600 in.
  • Tolerance of 0.000200 in.
  • Constructed from steel
  • Class ZZ precision
Catalog No. 50-005-11703 Supplier MSC Supplier No. 78375565
This Class ZZ minus plug and pin gage is designed for precise diameter measurements. Constructed from steel, it provides reliable performance in dimensional inspection and calibration applications.
  • Nominal diameter of 0.3600 in.
  • Tolerance of 0.000200 in.
  • Constructed from steel
  • Class ZZ precision