Learn More
MSC SPI Plug Gage, Class ZZ, Minus, Steel, 0.0370 in. Diameter, 1 ea.
Catalog No. 5000511727
Encompass
This Class ZZ minus plug and pin gage is designed for precise measurement and inspection applications. Constructed from durable steel, it provides reliable performance in metalworking and industrial environments.
  • Diameter of 0.0370 in.
  • Tolerance of 0.000200 in.
  • Constructed from steel
Catalog No. 50-005-11727 Supplier MSC Supplier No. 78372349
This Class ZZ minus plug and pin gage is designed for precise measurement and inspection applications. Constructed from durable steel, it provides reliable performance in metalworking and industrial environments.
  • Diameter of 0.0370 in.
  • Tolerance of 0.000200 in.
  • Constructed from steel