Learn More
MSC Swiss Precision Instruments Plug and Pin Gage, Steel, 0.854 in. Diameter, Class ZZ Plus, 1 ea.
Catalog No. 5000547717
Encompass
This Class ZZ Plus plug and pin gage is a precision measurement tool designed for checking hole sizes. It features a steel construction and is engineered to provide accurate diameter verification.
  • Diameter measurement of 0.854 in.
  • Class ZZ Plus precision rating
  • Tolerance of 0.000200 in.
  • Durable steel construction
Catalog No. 50-005-47717 Supplier MSC Supplier No. 78390390
Add to Cart
Add to Cart
This Class ZZ Plus plug and pin gage is a precision measurement tool designed for checking hole sizes. It features a steel construction and is engineered to provide accurate diameter verification.
  • Diameter measurement of 0.854 in.
  • Class ZZ Plus precision rating
  • Tolerance of 0.000200 in.
  • Durable steel construction