Learn More
MSC SPI Plug Gage, 0.764 in., Class ZZ, Steel, 1 ea.
Catalog No. 5000547718
Encompass
This Class ZZ plus plug gage is designed for precise measurement applications, featuring a diameter of 0.764 in. Manufactured for accuracy and durability, this tool is suitable for industrial inspection and quality control environments.
  • Diameter of 0.764 in.
  • Class ZZ plus tolerance
  • Tolerance of 0.000200 in.
  • Constructed from steel
  • Country of origin: USA
Catalog No. 50-005-47718 Supplier MSC Supplier No. 78389491
Add to Cart
Add to Cart
This Class ZZ plus plug gage is designed for precise measurement applications, featuring a diameter of 0.764 in. Manufactured for accuracy and durability, this tool is suitable for industrial inspection and quality control environments.
  • Diameter of 0.764 in.
  • Class ZZ plus tolerance
  • Tolerance of 0.000200 in.
  • Constructed from steel
  • Country of origin: USA