Learn More
MSC SPI Plug Gage, Steel, 0.5670 in. Diameter, 1 ea.
Catalog No. 5000547719
Encompass
This Class ZZ Plus plug gage is designed for precision inspection and measurement applications. Constructed from durable steel, it provides a reliable standard for go/no-go quality control tasks in metalworking and manufacturing environments.
  • Diameter of 0.5670 in.
  • Tolerance of 0.000200 in.
  • Steel construction for enhanced durability
Catalog No. 50-005-47719 Supplier MSC Supplier No. 78387529
This Class ZZ Plus plug gage is designed for precision inspection and measurement applications. Constructed from durable steel, it provides a reliable standard for go/no-go quality control tasks in metalworking and manufacturing environments.
  • Diameter of 0.5670 in.
  • Tolerance of 0.000200 in.
  • Steel construction for enhanced durability