Learn More
MSC SPI Class ZZ Minus Plug Gage, 0.0440 in. Diameter, Steel, 1 ea
Catalog No. 5000511682
Encompass
This Class ZZ minus plug and pin gage is a precision measurement tool designed for checking hole sizes, hole locations, and gaging slots. It is constructed from durable steel to ensure reliable performance during inspection tasks.
  • Class ZZ precision rating
  • Minus tolerance of 0.000200 in.
  • Diameter of 0.0440 in.
  • Designed for go/no-go inspection applications
  • Manufactured from high-quality steel
Catalog No. 50-005-11682 Supplier MSC Supplier No. 78372414
This Class ZZ minus plug and pin gage is a precision measurement tool designed for checking hole sizes, hole locations, and gaging slots. It is constructed from durable steel to ensure reliable performance during inspection tasks.
  • Class ZZ precision rating
  • Minus tolerance of 0.000200 in.
  • Diameter of 0.0440 in.
  • Designed for go/no-go inspection applications
  • Manufactured from high-quality steel