Learn More
MSC SPI Plug Gage, Class ZZ, Steel, 0.2580 in. Diameter, 1 ea.
Catalog No. 5000511685
Encompass
This Class ZZ minus plug and pin gage is a high-precision measurement tool designed for checking hole sizes, hole locations, and as a go/no-go gage for various machining and inspection applications. Engineered for durability, this gage provides accurate measurements with a consistent tolerance.
  • Constructed from high-quality steel for long-lasting wear resistance
  • Features a 0.2580 in. nominal diameter
  • Provides a tolerance of 0.0002 in.
  • Designed for precise dimensional inspection in metalworking and manufacturing environments
Catalog No. 50-005-11685 Supplier MSC Supplier No. 78374543
This Class ZZ minus plug and pin gage is a high-precision measurement tool designed for checking hole sizes, hole locations, and as a go/no-go gage for various machining and inspection applications. Engineered for durability, this gage provides accurate measurements with a consistent tolerance.
  • Constructed from high-quality steel for long-lasting wear resistance
  • Features a 0.2580 in. nominal diameter
  • Provides a tolerance of 0.0002 in.
  • Designed for precise dimensional inspection in metalworking and manufacturing environments