Learn More
MSC SPI Class ZZ Plus Plug Gage, Steel, 0.6120 in. Diameter, 1 ea
Catalog No. 5000511629
Encompass
This Class ZZ Plus plug gage is designed for precision inspection and measurement applications. It features a specific diameter of 0.6120 in. and is constructed from durable steel to ensure reliable performance in various metalworking and inspection environments.
  • Diameter: 0.6120 in.
  • Tolerance: 0.000200 in.
  • Material: Steel
  • Quantity: 1 per package
Catalog No. 50-005-11629 Supplier MSC Supplier No. 78387974
This Class ZZ Plus plug gage is designed for precision inspection and measurement applications. It features a specific diameter of 0.6120 in. and is constructed from durable steel to ensure reliable performance in various metalworking and inspection environments.
  • Diameter: 0.6120 in.
  • Tolerance: 0.000200 in.
  • Material: Steel
  • Quantity: 1 per package