Learn More
MSC SPI Plug Gage, Steel, 0.6320 in. Diameter, 1 ea.
Catalog No. 5000510937
Encompass
This Class ZZ Plus pin gage is a precision measuring tool designed for accurate hole diameter inspection and calibration. Manufactured from steel, this gage provides a diameter of 0.6320 in. with a tolerance of 0.000200 in.
  • Constructed from durable steel for long-lasting use
  • Class ZZ Plus precision rating
  • Diameter of 0.6320 in.
  • Tolerance of 0.000200 in.
  • Ideal for gauging hole sizes and inspection applications
Catalog No. 50-005-10937 Supplier MSC Supplier No. 78388170
This Class ZZ Plus pin gage is a precision measuring tool designed for accurate hole diameter inspection and calibration. Manufactured from steel, this gage provides a diameter of 0.6320 in. with a tolerance of 0.000200 in.
  • Constructed from durable steel for long-lasting use
  • Class ZZ Plus precision rating
  • Diameter of 0.6320 in.
  • Tolerance of 0.000200 in.
  • Ideal for gauging hole sizes and inspection applications