Learn More
MSC SPI Plug Gauge, Steel, 0.0002 in. Tolerance, 0.7400 in. Dia., 1 ea
Catalog No. 5000510938
Encompass
This Class ZZ Plus plug gage is designed for precision measurement applications, offering high-accuracy verification for hole sizing and tolerance checks. Constructed from durable steel, this gage is engineered to maintain consistency in demanding metrology environments.
  • Features a nominal diameter of 0.7400 in.
  • Tolerance rating of 0.0002 in.
  • Constructed from steel for long-term wear resistance
  • Reliable performance for precision inspection tasks
Catalog No. 50-005-10938 Supplier MSC Supplier No. 78389251
This Class ZZ Plus plug gage is designed for precision measurement applications, offering high-accuracy verification for hole sizing and tolerance checks. Constructed from durable steel, this gage is engineered to maintain consistency in demanding metrology environments.
  • Features a nominal diameter of 0.7400 in.
  • Tolerance rating of 0.0002 in.
  • Constructed from steel for long-term wear resistance
  • Reliable performance for precision inspection tasks