Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.991 in. Diameter, 0.0002 in. Tolerance, 1 ea.
Catalog No. 5000510917
Encompass
This Class ZZ plus plug gage is a precision measurement tool manufactured from steel, designed for reliable and accurate inspection of bore diameters.
  • Diameter of 0.9910 in.
  • Tolerance of 0.0002 in.
  • Constructed from steel
  • Manufactured in the USA
Catalog No. 50-005-10917 Supplier MSC Supplier No. 78391760
This Class ZZ plus plug gage is a precision measurement tool manufactured from steel, designed for reliable and accurate inspection of bore diameters.
  • Diameter of 0.9910 in.
  • Tolerance of 0.0002 in.
  • Constructed from steel
  • Manufactured in the USA