Learn More
MSC SPI Class ZZ Minus Plug Gage, Steel, 0.2110 in. Diameter, 1 ea.
Catalog No. 5000510137
Encompass
This Class ZZ plug gage is designed for precise diameter measurements and calibration tasks. Engineered from durable steel, this gage provides reliable performance in industrial inspection environments.
  • Constructed from high-quality steel
  • Diameter of 0.2110 in.
  • Tolerance of 0.000200 in.
  • Class ZZ accuracy
Catalog No. 50-005-10137 Supplier MSC Supplier No. 78374071
This Class ZZ plug gage is designed for precise diameter measurements and calibration tasks. Engineered from durable steel, this gage provides reliable performance in industrial inspection environments.
  • Constructed from high-quality steel
  • Diameter of 0.2110 in.
  • Tolerance of 0.000200 in.
  • Class ZZ accuracy