Learn More
MSC SPI Class ZZ Minus Plug Gage, Steel, 0.7210 in. Diameter, 1 ea.
Catalog No. 5000510125
Encompass
This Class ZZ minus plug and pin gage is designed for precise measurement and inspection applications. Featuring a nominal diameter of 0.7210 in., it provides reliable accuracy for checking hole sizes, hole locations, and slot widths in manufacturing and machining environments.
  • Nominal diameter of 0.7210 in.
  • Constructed from durable steel
  • Class ZZ tolerance of 0.000200 in.
  • Minus tolerance designation
Catalog No. 50-005-10125 Supplier MSC Supplier No. 78379179
This Class ZZ minus plug and pin gage is designed for precise measurement and inspection applications. Featuring a nominal diameter of 0.7210 in., it provides reliable accuracy for checking hole sizes, hole locations, and slot widths in manufacturing and machining environments.
  • Nominal diameter of 0.7210 in.
  • Constructed from durable steel
  • Class ZZ tolerance of 0.000200 in.
  • Minus tolerance designation