Learn More
MSC SPI Plug Gage, Class ZZ Minus, Steel, 0.9010 in., 1 ea
Catalog No. 5000510123
Encompass
This Class ZZ plug and pin gage is designed for precise measurement and inspection applications. Constructed from durable steel, it provides a reliable standard for checking hole sizes and tolerances in various machining and quality control environments.
  • Nominal diameter of 0.9010 in.
  • Class ZZ accuracy with a tolerance of 0.0002 in.
  • Constructed from steel for durability
  • Designed for decimal inch measurement systems
Catalog No. 50-005-10123 Supplier MSC Supplier No. 78380979
This Class ZZ plug and pin gage is designed for precise measurement and inspection applications. Constructed from durable steel, it provides a reliable standard for checking hole sizes and tolerances in various machining and quality control environments.
  • Nominal diameter of 0.9010 in.
  • Class ZZ accuracy with a tolerance of 0.0002 in.
  • Constructed from steel for durability
  • Designed for decimal inch measurement systems