Learn More
MSC SPI Class ZZ Minus Plug Gage, Steel, 0.9600 in. Diameter, 1 ea
Catalog No. 5000516416
Encompass
This Class ZZ minus plug gage is designed for precise measurement applications, featuring a diameter of 0.9600 in. Constructed from durable steel, it is suitable for a variety of inspection tasks in industrial environments.
  • Diameter of measurement: 0.9600 in.
  • Tolerance: 0.000200 in.
  • Material: Steel
  • System of measurement: Decimal inch
Catalog No. 50-005-16416 Supplier MSC Supplier No. 78381563
This Class ZZ minus plug gage is designed for precise measurement applications, featuring a diameter of 0.9600 in. Constructed from durable steel, it is suitable for a variety of inspection tasks in industrial environments.
  • Diameter of measurement: 0.9600 in.
  • Tolerance: 0.000200 in.
  • Material: Steel
  • System of measurement: Decimal inch