Learn More
MSC SPI Class ZZ Plus Plug Gage, Steel, 0.2530 in. Diameter, 1 ea.
Catalog No. 5000516418
Encompass
The Class ZZ Plus plug gage is a precision measurement tool designed for verifying hole sizes with high accuracy. This gage is constructed from durable steel and provides a reliable solution for quality control and inspection tasks in metalworking and manufacturing environments.
  • Diameter: 0.2530 in.
  • Tolerance: 0.000200 in.
  • Material: Steel
  • Country of origin: China
Catalog No. 50-005-16418 Supplier MSC Supplier No. 78384385
The Class ZZ Plus plug gage is a precision measurement tool designed for verifying hole sizes with high accuracy. This gage is constructed from durable steel and provides a reliable solution for quality control and inspection tasks in metalworking and manufacturing environments.
  • Diameter: 0.2530 in.
  • Tolerance: 0.000200 in.
  • Material: Steel
  • Country of origin: China