Learn More
MSC SPI Plug and Pin Gage, Class ZZ, Minus, 0.9090 in. Diameter, 1 ea.
Catalog No. 5000516423
Encompass
This Class ZZ plug and pin gage is designed for precision measurement applications in industrial and machining environments. Featuring a 0.9090 in. diameter, this tool provides reliable reference for calibration and inspection tasks.
  • Constructed from durable steel
  • Precision diameter of 0.9090 in.
  • Tolerance of 0.000200 in.
  • Decimal inch measurement system
  • Manufactured in the USA
Catalog No. 50-005-16423 Supplier MSC Supplier No. 78381050
This Class ZZ plug and pin gage is designed for precision measurement applications in industrial and machining environments. Featuring a 0.9090 in. diameter, this tool provides reliable reference for calibration and inspection tasks.
  • Constructed from durable steel
  • Precision diameter of 0.9090 in.
  • Tolerance of 0.000200 in.
  • Decimal inch measurement system
  • Manufactured in the USA