Learn More
MSC SPI Plug Gage, Steel, 0.6330 in. Diameter, 0.000200 in. Tolerance, 1 ea
Catalog No. 5000516394
Encompass
This Class ZZ minus plug gage is a precision measurement tool designed for accurate hole diameter inspection and calibration tasks. Manufactured from steel, this gage provides durable performance for metalworking and quality control applications.
  • Diameter of 0.6330 in.
  • Class ZZ tolerance of 0.000200 in.
  • Constructed from steel
  • Decimal inch system of measurement
Catalog No. 50-005-16394 Supplier MSC Supplier No. 78378296
This Class ZZ minus plug gage is a precision measurement tool designed for accurate hole diameter inspection and calibration tasks. Manufactured from steel, this gage provides durable performance for metalworking and quality control applications.
  • Diameter of 0.6330 in.
  • Class ZZ tolerance of 0.000200 in.
  • Constructed from steel
  • Decimal inch system of measurement