Learn More
MSC SPI Swiss Precision Instruments Class ZZ Plug Gage, Steel, 0.859 in. Diameter, 0.0002 in. Tolerance, 1 ea.
Catalog No. 5000516396
Encompass
This Class ZZ minus plug and pin gage is designed for high-precision measurement applications. Manufactured from high-quality steel, it provides a diameter of 0.8590 in. with a tolerance of 0.000200 in.
  • Constructed from durable steel for long-lasting performance
  • Designed for precise diameter measurements
  • Features a 0.8590 in. diameter
  • Maintains a 0.000200 in. tolerance
  • Originates from the USA
Catalog No. 50-005-16396 Supplier MSC Supplier No. 78380557
This Class ZZ minus plug and pin gage is designed for high-precision measurement applications. Manufactured from high-quality steel, it provides a diameter of 0.8590 in. with a tolerance of 0.000200 in.
  • Constructed from durable steel for long-lasting performance
  • Designed for precise diameter measurements
  • Features a 0.8590 in. diameter
  • Maintains a 0.000200 in. tolerance
  • Originates from the USA