Learn More
MSC SPI Plug and Pin Gage, Class ZZ Minus, 0.7910 in. Diameter, Steel, 1 ea.
Catalog No. 5000516409
Encompass
This Class ZZ minus plug and pin gage is designed for precision inspection and measurement applications. Constructed from durable steel, this gage provides a reliable standard for go/no-go testing in metalworking and manufacturing environments.
  • Class ZZ minus tolerance
  • Diameter of 0.7910 in.
  • Tolerance of 0.000200 in.
  • Constructed from steel
  • Decimal inch measurement system
Catalog No. 50-005-16409 Supplier MSC Supplier No. 78379872
This Class ZZ minus plug and pin gage is designed for precision inspection and measurement applications. Constructed from durable steel, this gage provides a reliable standard for go/no-go testing in metalworking and manufacturing environments.
  • Class ZZ minus tolerance
  • Diameter of 0.7910 in.
  • Tolerance of 0.000200 in.
  • Constructed from steel
  • Decimal inch measurement system