Learn More
MSC SPI Class ZZ Plus Plug Gage, Steel, 0.5090 in. Diameter, 0.0002 in. Tolerance, 1 ea
Catalog No. 5000516364
Encompass
This Class ZZ Plus plug gage is designed for precise measurement applications, featuring a 0.5090 in. diameter. Constructed from durable steel, this gage ensures reliable performance in industrial inspecting and testing environments.
  • Gauge diameter: 0.5090 in.
  • Tolerance: 0.0002 in.
  • Material: Steel
Catalog No. 50-005-16364 Supplier MSC Supplier No. 78386943
This Class ZZ Plus plug gage is designed for precise measurement applications, featuring a 0.5090 in. diameter. Constructed from durable steel, this gage ensures reliable performance in industrial inspecting and testing environments.
  • Gauge diameter: 0.5090 in.
  • Tolerance: 0.0002 in.
  • Material: Steel