Learn More
MSC SPI Class ZZ Plus Plug Gage, Steel, 0.1560 in. Diameter, 1 ea.
Catalog No. 5000516108
Encompass
This Class ZZ Plus plug gage is designed for precise measurement and inspection applications. Constructed from durable steel, this gage provides accurate diameter verification for quality control tasks in metalworking and manufacturing environments.
  • Diameter of 0.1560 in.
  • Tolerance of 0.000200 in.
  • Constructed from steel
Catalog No. 50-005-16108 Supplier MSC Supplier No. 78383411
This Class ZZ Plus plug gage is designed for precise measurement and inspection applications. Constructed from durable steel, this gage provides accurate diameter verification for quality control tasks in metalworking and manufacturing environments.
  • Diameter of 0.1560 in.
  • Tolerance of 0.000200 in.
  • Constructed from steel