Learn More
MSC SPI Class ZZ Plus Plug Gage, Steel, 0.3020 in. Diameter, 1 ea.
Catalog No. 5000516109
Encompass
This Class ZZ Plus plug gage is designed for precision measurement and inspection applications. It features a nominal diameter of 0.3020 in. and is constructed from steel for durability in demanding industrial environments.
  • Nominal diameter of 0.3020 in.
  • Tolerance of 0.000200 in.
  • Constructed from steel
Catalog No. 50-005-16109 Supplier MSC Supplier No. 78384872
This Class ZZ Plus plug gage is designed for precision measurement and inspection applications. It features a nominal diameter of 0.3020 in. and is constructed from steel for durability in demanding industrial environments.
  • Nominal diameter of 0.3020 in.
  • Tolerance of 0.000200 in.
  • Constructed from steel