Learn More
MSC SPI Plug Gage, Class ZZ, Steel, 0.9680 in. Diameter, 1 ea.
Catalog No. 5000515467
Encompass
This Class ZZ minus plug gage is designed for precise diameter measurement and inspection applications. Constructed from durable steel, this gage provides reliable performance in industrial metrology environments.
  • Diameter of measurement: 0.9680 in.
  • Tolerance: 0.000200 in.
  • System of measurement: Decimal inch
  • Material: Steel
Catalog No. 50-005-15467 Supplier MSC Supplier No. 78381647
This Class ZZ minus plug gage is designed for precise diameter measurement and inspection applications. Constructed from durable steel, this gage provides reliable performance in industrial metrology environments.
  • Diameter of measurement: 0.9680 in.
  • Tolerance: 0.000200 in.
  • System of measurement: Decimal inch
  • Material: Steel