Learn More
MSC SPI Class ZZ Plus Plug Gage, Steel, 0.3750 in. Diameter, 1 ea
Catalog No. 5000515429
Encompass
This Class ZZ Plus plug gage is a precision measuring tool designed for accurate inspection of hole diameters. Constructed from durable steel, it provides a reliable standard for go/no go gauging applications in metalworking and manufacturing environments.
  • Diameter of 0.3750 in.
  • Tolerance of 0.0002 in.
  • Constructed from steel
Catalog No. 50-005-15429 Supplier MSC Supplier No. 78385606
This Class ZZ Plus plug gage is a precision measuring tool designed for accurate inspection of hole diameters. Constructed from durable steel, it provides a reliable standard for go/no go gauging applications in metalworking and manufacturing environments.
  • Diameter of 0.3750 in.
  • Tolerance of 0.0002 in.
  • Constructed from steel