Learn More
MSC SPI Swiss Precision Instruments Plug Gage, 0.782 in. Diameter, 0.0002 in. Tolerance, Steel, 1 ea.
Catalog No. 5000515184
Encompass
This Class ZZ plus plug gage is manufactured from steel and designed for high-precision measurement applications. It provides a nominal diameter of 0.782 in. with a tolerance of 0.0002 in.
  • Constructed from steel for durability
  • Designed for accurate gaging requirements
  • Meets Class ZZ specifications
Catalog No. 50-005-15184 Supplier MSC Supplier No. 78389673
This Class ZZ plus plug gage is manufactured from steel and designed for high-precision measurement applications. It provides a nominal diameter of 0.782 in. with a tolerance of 0.0002 in.
  • Constructed from steel for durability
  • Designed for accurate gaging requirements
  • Meets Class ZZ specifications