Learn More
MSC SPI Class ZZ Plus Plug Gage, Steel, 0.5780 in. Diameter, 1 ea.
Catalog No. 5000515128
Encompass
This Class ZZ plus plug gage is a high-precision measurement tool designed for reliable inspection and calibration applications. Constructed from durable steel, it is engineered for use in Go inspection, setting micrometers, checking verniers, and calibrating other precision measuring devices.
  • Diameter of 0.5780 in.
  • Tolerance of 0.000200 in.
  • Manufactured from robust steel for long-term wear resistance
  • Suitable for Go inspection tasks
  • Ideal for setting and calibrating micrometers and verniers
Catalog No. 50-005-15128 Supplier MSC Supplier No. 78387636
This Class ZZ plus plug gage is a high-precision measurement tool designed for reliable inspection and calibration applications. Constructed from durable steel, it is engineered for use in Go inspection, setting micrometers, checking verniers, and calibrating other precision measuring devices.
  • Diameter of 0.5780 in.
  • Tolerance of 0.000200 in.
  • Manufactured from robust steel for long-term wear resistance
  • Suitable for Go inspection tasks
  • Ideal for setting and calibrating micrometers and verniers